missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIC4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00172
This item is not returnable.
View return policy
Description
CLIC4 Polyclonal specifically detects CLIC4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| CLIC4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| chloride intracellular channel 4, chloride intracellular channel 4 like, chloride intracellular channel protein 4, CLIC4L, DKFZp566G223, FLJ38640, H1, huH1, Intracellular chloride ion channel protein p64H1, MTCLIC, p64H1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25932 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1-0.3 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA detection limit 1:64000 | |
| NP_039234.1 | |
| CLIC4 | |
| Peptide with sequence NGLKEEDKEPLIE-C corresponding to N-Terminus according to NP_039234.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected to react with Rat and Dog based on Sequence similarity. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction