missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIC4 Antibody (JE65-09), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32192
This item is not returnable.
View return policy
Description
CLIC4 Monoclonal antibody specifically detects CLIC4 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CLIC4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:200 | |
| chloride intracellular channel 4, chloride intracellular channel 4 like, chloride intracellular channel protein 4, CLIC4L, DKFZp566G223, FLJ38640, H1, huH1, Intracellular chloride ion channel protein p64H1, MTCLIC, p64H1 | |
| Synthetic peptide. (Uniprot: Q9Y696) | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction | |
| 25932 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| JE65-09 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction