missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLIC3 Monoclonal Antibody (2C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520574
This item is not returnable.
View return policy
Description
Peptide Sequence: RLDSYLRAPL EHELAGEPQL RESRRRFLDG DRLTLADCSL LPKLHIVDTV CAHFRQAPIP AELRGVRRYL DSAMQEKEFK YTCPHSAEIL AAYRPAVHPR.
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 3 is a member of the p64 family and is predominantly localized in the nucleus and stimulates chloride ion channel activity. In addition, this protein may participate in cellular growth control, based on its association with ERK7, a member of the MAP kinase family.
Specifications
| CLIC3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O95833 | |
| CLIC3 | |
| CLIC3 (NP_004660, 137 a.a. ~ 236 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2C5 | |
| Unconjugated | |
| CLIC3 | |
| 2300003G24Rik; chloride intracellular channel 3; chloride intracellular channel protein 3; CLIC3; RP11-229P13.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9022 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction