missing translation for 'onlineSavingsMsg'
Learn More

CLIC1 Antibody, Novus Biologicals™

Product Code. 18396097 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396097 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396097 Supplier Novus Biologicals Supplier No. H00001192B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CLIC1 Polyclonal antibody specifically detects CLIC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 to 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 to 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_001279.2
Gene Alias Chloride channel ABP, chloride intracellular channel 1, chloride intracellular channel protein 1, G6, hRNCC, NCC27p64CLCP, Nuclear chloride ion channel 27, nuclear chloride ion channel protein, Regulatory nuclear chloride ion channel protein, RNCC protein
Host Species Mouse
Immunogen CLIC1 (NP_001279.2, 1 a.a. - 241 a.a.) full-length human protein. MAEEQPQVELFVKAGSDGAKIGNCPFSQRLFMVLWLKGVTFNVTTVDTKRRTETVQKLCPGGQLPFLLYGTEVHTDTNKIEEFLEAVLCPPRYPKLAALNPESNTAGLDIFAKFSAYIKNSNPALNDNLEKGLLKALKVLDNYLTSPLPEEVDETSAEDEGVSQRKFLDGNELTLADCNLLPKLHIVQVVCKKYRGFTIPEAFRGVHRYLSNAYAREEFASTCPDDEEIELAYEQVAKALK
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1192
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.