missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561815
This item is not returnable.
View return policy
Description
Immunogen sequence: ESEPEEKSEE EIEIIEGQEE SNQSNKSGSE DEMKEADEST GSGDGPIKSV RKR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 74%.
Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.
Specifications
| CLGN | |
| Polyclonal | |
| Unconjugated | |
| CLGN | |
| 4930459O04Rik; A2/6; AI528775; Calmegin; calnexin-T; Clgn; Cln; hypothetical protein LOC504256; LOW QUALITY PROTEIN: calmegin; MEG 1 antigen; Meg1; testis tissue sperm-binding protein Li 79P | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1047 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14967 | |
| CLGN | |
| Recombinant protein corresponding to Human CLGN. Recombinant protein control fragment (Product #RP-103753). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction