missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clenbuterol Antibody (PSH0-01), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32191
This item is not returnable.
View return policy
Description
Clenbuterol Monoclonal antibody specifically detects Clenbuterol in Nonspecies specific samples. It is validated for ELISA
Specifications
| Clenbuterol | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:5000-1:20000 | |
| 1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol, 37148-27-9, 4-Amino-3,5-dichloro-alpha-(((1,1-dimethylethyl)amino)methyl)benzenemethanol, Clenbuterolum, NAB 365, Planipart | |
| Clenbuterol-OVA | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| PSH0-01 | |
| Unconjugated | |
| PBS (pH7.4) | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction