missing translation for 'onlineSavingsMsg'
Learn More

Clemizole hydrochloride, MedChemExpress

Product Code. 16448028
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16448028

Brand: Medchem Express HY30234A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clemizole hydrochloride is an H1 histamine receptor antagonist, is found to substantially inhibit HCV replication. Clemizole hydrochloride is an inhibitor of TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, whereas its EC50 for viral replication is 8 μM.

Clemizole hydrochloride is an H1 histamine receptor antagonist, is found to substantially inhibit HCV replication. Clemizole hydrochloride is an inhibitor of TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, whereas its EC50 for viral replication is 8 μM.

Specifications

Chemical Name or Material Clemizole hydrochloride
CAS 1163-36-6
Color White
Purity Grade Notes Research
Molecular Formula C19H21Cl2N3
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (92.00 mM; Need ultrasonic) ∣H2O : 20 mg/mL (55.20 mM; Need ultrasonic)
SMILES ClC1=CC=C(C=C1)CN2C(CN3CCCC3)=NC4=CC=CC=C24.[H]Cl
Molecular Weight (g/mol) 362.3
Formula Weight 362.3
Percent Purity 99.24%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.