missing translation for 'onlineSavingsMsg'
Learn More

Clemastine N-Oxide, TRC

Product Code. 30133596
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133596 100 mg 100mg
30012152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133596 Supplier Toronto Research Chemicals Supplier No. TRCC568505100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108825-05-4
Molecular Formula C21 H26 Cl N O2
Molecular Weight (g/mol) 359.89
Synonym Pyrrolidine, 2-[2-[1-(4-chlorophenyl)-1-phenylethoxy]ethyl]-1-methyl-, 1-oxide, [R-(R*,R*)]- (9CI), (1RS,2R)-2-[2-[(1R)-1-(4-Chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine 1-Oxide, (1RS,2R)-2-[2-[(1R)-1-(4-Chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine 1-Oxide, [R-(R*,R*)]- 2-[2-[1-(4-Chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine 1-Oxide, Clemastine EP Impurity A
IUPAC Name (2R)-2-[2-[(1R)-1-(4-chlorophenyl)-1-phenyl-ethoxy]ethyl]-1-methyl-1-oxido-pyrrolidin-1-ium
SMILES C[C@@](OCC[C@H]1CCC[N+]1(C)[O-])(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H26ClNO2/c1-21(17-7-4-3-5-8-17,18-10-12-19(22)13-11-18)25-16-14-20-9-6-15-23(20,2)24/h3-5,7-8,10-13,20H,6,9,14-16H2,1-2H3/t20-,21-,23?/m1/s1
Formula Weight 359.1652
Chemical Name or Material Clemastine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.