missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC9A Recombinant Human Monoclonal Antibody (9H7)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560831
This item is not returnable.
View return policy
Description
CLEC9A Recombinant Monoclonal Antibody for ELISA
C-type lectin domain family 9 member A (CLEC9A) is a gene located on chromosome 12p13.2, encoding a protein that belongs to the C-type lectin receptor family. CLEC9A is primarily expressed on dendritic cells and certain subsets of macrophages, playing a pivotal role in the immune system by recognizing and binding dead cell debris, including necrotic cells. This receptor facilitates the presentation of antigens derived from dead cells to T cells, thus contributing to the initiation of immune responses. CLEC9A is involved in cross-presentation, a process crucial for adaptive immunity, particularly in the context of viral infections and cancer. Its expression and activity are of particular interest for vaccine development and immunotherapy, given its ability to enhance antigen presentation and stimulate robust immune responses. Research into CLEC9A continues to explore its therapeutic potential for modulating immune activity in various diseases, including cancer and infectious diseases.
Specifications
| CLEC9A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6UXN8 | |
| CLEC9A | |
| Recombinant Human CLEC9A protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA | |
| 9H7 | |
| Unconjugated | |
| CLEC9A | |
| 9830005G06Rik; CD370; Clec9a; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; Dendritic cell natural killer lectin group receptor 1; DNGR1; DNGR-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046 | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 283420 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction