missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55451-1-AP-20UL
This item is not returnable.
View return policy
Description
CLEC9A is a group V CTLR (C-type lectin-like receptor) that functions as an activation receptor and is expressed on myeloid lineage cells. This protein is also expressed at the cell surface and acts as glycosylated dimers which mediate endocytosis, but not phagocytosis. It facilitates antigen uptake and presentation, and may provide a suitable target for antibody-mediated antigen delivery. CLEC9A is important for conveying information from necrotic cells to T-cells and defines a pathway by which adaptive immune responses can be initiated in the absence of infection. It recruits Syk to mediate sensing of necrosis by the principal DC subset involved in regulating cross priming to cell-associated antigens. Signaling via this kinase occurs through a novel pathway involving CARD9 which induce a variety of cellular responses including the induction of cytokines (such as TNF, IL-6, IL-10, IL-23, and IL-2), the respiratory burst, and the production of Arachidonic acid. In addition, it also induces TH17 (T-helper type 17) adaptive responses in vivo. Expression of CLEC9A is highly restricted in peripheral blood, being detected only on BDCA3+ DCs and on a small subset of CD14+ CD16- monocytes. CLEC9A serves as a new marker to distinguish subtypes of DCs.Specifications
| CLEC9A | |
| Polyclonal | |
| Unconjugated | |
| CLEC9A | |
| CD370, CLEC9A, DNGR1, UNQ9341 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283420 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXN8 | |
| CLEC9A | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction