missing translation for 'onlineSavingsMsg'
Learn More

CLEC9a Antibody (14N8D7), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18401626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401626 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401626 Supplier Novus Biologicals Supplier No. NBP227087JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC9a Monoclonal antibody specifically detects CLEC9a in Human, Primate, Macaque samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC9a
Applications Western Blot
Classification Monoclonal
Clone 14N8D7
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Host Species Mouse
Immunogen A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Human, Primate, Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.