Learn More
Invitrogen™ CLEC5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114187
Description
Immunogen sequence: NTPEKLKFLQ DITDAEKYFI GLIYHREEKR WRWINNSVFN GNVTNQNQNF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Horse: 86%; Human: 100%; Pig: 79%; Rat: 79%; Yeast: 83%.
Functions as a positive regulator of osteoclastogenesis. Cell surface receptor that signals via TYROBP. Regulates inflammatory responses. Acts as a key regulator of synovial injury and bone erosion during autoimmune joint inflammation. Critical macrophage receptor for dengue virus serotypes 1-4. The binding of dengue virus to CLEC5A triggers signaling through the phosphylation of TYROBP, this interaction does not result in viral entry, but stimulates proinflammatory cytokine release.
Specifications
| CLEC5A | |
| Polyclonal | |
| Unconjugated | |
| CLEC5A | |
| CLEC5A; Clecsf5; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 5; C-type lectin domain family 5 member A; C-type lectin domain family 5, member a; C-type lectin superfamily member 5; Ly100; Mdl1; MDL-1; myeloid DAP12-associating lectin 1; myeloid DAP12-associating lectin-1; type II transmembrane protein MDL-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23601 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NY25 | |
| CLEC5A | |
| Immunogen sequence: NTPEKLKFLQ DITDAEKYFI GLIYHREEKR WRWINNSVFN GNVTNQNQNF (aa 110-159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.