missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4M, anti-Human, Clone: 16E7, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00096.200ug
This item is not returnable.
View return policy
Specifications
| CLEC4M | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| CD209L/CD299/DC-SIGN2/DC-SIGNR/DCSIGNR/HP10347/L-SIGN/LSIGN/MGC129964/MGC47866 | |
| Original antibody is raised against human CD299. | |
| RUO | |
| 10332 | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry | |
| 16E7 | |
| Flow Cytometry (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| C-type lectin domain family 4, member M | |
| CLEC4M | |
| 200 μg | |
| Primary | |
| This antibody recognizes human CLEC4M, which is an oligomeric type II transmembrane protein with a C-type lectin extracellular domain, the expression of which is restricted to immature DC, macrophages in the lung, and endothelial cells in the liver. It binds ICAM-3 and ICAM-7 to mediate the interaction of DC with T lymphocytes and endothelial cells in the initial stages of immune response and in the migratory behavior of DC. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction