missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC4F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548162
This item is not returnable.
View return policy
Description
CLEC4F Polyclonal Antibody for Western Blot, IHC (P)
Receptor with an affinity for galactose and fucose. Could be involved in endocytosis. Target protein's carbohydrate binding receptor with an affinity for fucose and galactose.
Specifications
| CLEC4F | |
| Polyclonal | |
| Unconjugated | |
| Clec4f | |
| Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 165530 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8N1N0 | |
| Clec4f | |
| KLH conjugated synthetic peptide between 311-339 amino acids from the C-terminal region of human CLEC4F. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction