missing translation for 'onlineSavingsMsg'
Learn More

CLEC4E Antibody (WEN43), Novus Biologicals™

Product Code. 18324313 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324313 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324313 Supplier Novus Biologicals Supplier No. NBP311192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC4E Monoclonal specifically detects CLEC4E in Rat samples. It is validated for Flow Cytometry, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4E
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone WEN43
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9
Host Species Mouse
Immunogen Rat CLEC4E-Fc gamma 2b fusion protein
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26253
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.