missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4E Antibody (16E3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-49311PEATT594
This item is not returnable.
View return policy
Description
CLEC4E Monoclonal specifically detects CLEC4E in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| CLEC4E | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CLEC4E | |
| Recombinant human CLEC4E (41-219aa) purified from E. coli (NP_055173). | |
| 0.1 mL | |
| Primary | |
| Human CLEC4E (41-219aa) | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 16000 | |
| Flow Cytometry | |
| CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
| Mouse | |
| Protein G purified | |
| Immunology | |
| 26253.0 | |
| Human, Rat | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction