missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CLEC4D/CLECSF8 Antibody Pair [HRP]

Product Code. 18208317 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18208317

Brand: Novus Biologicals™ NBP27963815PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human CLEC4D/CLECSF8.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 24 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human CLEC4D/CLECSF8.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL
Gene ID (Entrez) 338339
Gene Symbol CLEC4D
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 80 ng of recombinant CLEC4D/CLECSF8.
  • Capture Antibody: 0.3 mg/mL of mouse anti-CLEC4D/CLECSF8 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-CLEC4D/CLECSF8 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target CLEC4D/CLECSF8
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.