missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC3B/Tetranectin Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00044CL1
This item is not returnable.
View return policy
Description
CLEC3B/Tetranectin Polyclonal antibody specifically detects CLEC3B/Tetranectin in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| CLEC3B/Tetranectin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
| Produced in rabbits immunized with purified, recombinant Human CLEC3B/Tetranectin (Accession#: AAX37102.1; Met 1-Val 202) | |
| 0.1 mL | |
| Cancer | |
| 7123 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction