missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC3B/Tetranectin Antibody (011), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CLEC3B/Tetranectin Monoclonal antibody specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Gene Alias | C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin |
| Host Species | Rabbit |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?