missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570780
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%.
CLEC3A gene ontology annotations related to this gene include carbohydrate binding.
Specifications
| CLEC3A | |
| Polyclonal | |
| Unconjugated | |
| CLEC3A | |
| 1110019O10Rik; Cartilage-derived C-type lectin; clec3a; CLECSF1; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 1 (cartilage-derived); C-type calcium dependent, carbohydrate-recognition domain lectin, superfamily member 1, cartilage-derived; C-type lectin domain family 3 member A; C-type lectin domain family 3, member a; C-type lectin homolog precursor; C-type lectin superfamily member 1; domain) lectin, superfamily member 1(cartilage-derived); Gm796; UNQ700/PRO1345 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10143 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75596 | |
| CLEC3A | |
| synthetic peptide directed towards the C-terminal region of human CLEC3A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction