missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC2D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553617
This item is not returnable.
View return policy
Description
Immunogen sequence: RANCHQEPSV CLQAACPESW IGFQRKCFYF SDDTKNWTSS QRFCDSQDAD LAQVESFQEL NFLLRYKGPS DHWIGLSREQ GQPWKWINGT EWTRQFPILG A Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.
Recently, Zhou et al have identified a protein that can inhibit multinucleate Osteoclast formation in murine osteoblast and spleen cell co-cultures. This protein called, Osteoclast Inhibitory Lectin (OCIL) can also inhibit osteoclast formation in spleen cell cultures treated with RANKL and M-CSF. Furthermore, mOCIL acted directly on macrophage/monocyte cells as evidenced by its inhibitory action on adherent spleen cell cultures, which were depleted of stromal and lymphocytic cells. mOCIL completely inhibited osteoclast formation during the proliferative phase of osteoclast formation and resulted in 70% inhibition during the differentiation phase. Osteoblast OCIL mRNA expression was enhanced by parathyroid hormone, calcitriol, interleukin-1 and IL-11, and retinoic acid. OCIL is expressed in osteoblasts and chondrocytes as well as in a variety of extraskeletal tissues. Since OCIL expression was found to be very similar to distribution for RANKL, it has been suggested that RANKl and OCIL may interact in skeleton and in extracellular tissues.
Specifications
| CLEC2D | |
| Polyclonal | |
| Unconjugated | |
| CLEC2D | |
| CLAX; Clec2d; Clrb; Clr-b; C-type lectin domain family 2 member D; C-type lectin domain family 2, member d; C-type lectin related f; C-type lectin superfamily 2, member D; C-type lectin-domain family 2 member D; C-type lectin-related protein B; lectin-like NK cell receptor; Lectin-like transcript 1; lectin-like transmembrane protein; LLT1; LLT-1; MGC123478; OCIL; Osteoclast inhibitory lectin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHP7 | |
| CLEC2D | |
| Recombinant protein corresponding to Human CLEC2D. Recombinant protein control fragment (Product #RP-89969). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction