missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC1B Monoclonal Antibody (AYP1), Biotin

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528529
This item is not returnable.
View return policy
Description
CLEC1B Monoclonal Antibody for ICC/IF, Flow, IP
Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
Specifications
| CLEC1B | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide; pH 7.4 | |
| Q9P126 | |
| CLEC1B | |
| A recombinant extracellular fragment of human CLEC-2 (aa 68-229 extracellular domain). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| AYP1 | |
| Biotin | |
| CLEC1B | |
| 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384 | |
| Mouse | |
| Protein G | |
| RUO | |
| 51266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction