missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC2A Monoclonal Antibody (705518)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561783
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
CLEC2A (C-type lectin domain family 2 member A), also known as PILAR (proliferation-induced lymphocyte-associated receptor) is involved in modulating T-cell expansion. It is a 32 kDa single-pass type II membrane protein that contains one C-type lectin domain in its extracellular region and belongs to the CTL/CTLD superfamily. CLEC2A is mainly expressed in skin and highly expressed in CD8(+), B lymphocytes and naive CD4(+) T cells. Manipulation of CLEC2A signaling may be important for treatment of autoimmune diseases and cancer.
Specifications
| CLEC2A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UVW9 | |
| CLEC2A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC-2A, Trp49-Leu174 (predicted). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 705518 | |
| Unconjugated | |
| CLEC2A | |
| CLEC2A; C-type lectin domain family 2 member A; C-type lectin domain family 2, member A; INPE5792; KACL; Keratinocyte-associated C-type lectin; PILAR; Proliferation-induced lymphocyte-associated receptor; UNQ579; UNQ5792; UNQ5792/PRO19597 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 387836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction