missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13394-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded protein may play a role in regulating dendritic cell function. Alternative splice variants have been described but their biological nature has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.Specifications
| CLEC1A | |
| Polyclonal | |
| Unconjugated | |
| CLEC1A | |
| C type lectin like receptor 1, CLEC 1, CLEC1, CLEC1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 243653, 500337, 51267 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BWY2, Q8NC01 | |
| CLEC1A | |
| CLEC1A Fusion Protein Ag4058 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction