missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC18A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21013-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds polysaccharides in a Ca(2+)-independent manner with a preferentially binding to fucoidan, beta-glucans and galactans.Specifications
| CLEC18A | |
| Polyclonal | |
| Unconjugated | |
| CLEC18A | |
| CLEC18A, MRCL, MRCL1, MRLP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 307851, 348174, 353287 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A5D8T8, Q7TSQ1 | |
| CLEC18A | |
| CLEC18A Fusion Protein Ag15237 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction