missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC16A Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00171
This item is not returnable.
View return policy
Description
CLEC16A Polyclonal specifically detects CLEC16A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| CLEC16A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C-type lectin domain family 16, member A, KIAA0350Gop-1, MGC111457, protein CLEC16A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_056041.1 | |
| CLEC16A | |
| Peptide with sequence C-SLENQDKGGERP corresponding to internal region according to NP_056041.1. | |
| 0.1 mg | |
| Immunology, Innate Immunity | |
| 23274 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction