missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC14A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CLEC14A |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882555
|
Proteintech
23964-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872555
|
Proteintech
23964-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC-14A (C-type lectin domain family 14 member A), also known as EGFR-5 (epidermal growth factor receptor 5) or CEG1, is a 490 amino acid single-pass type I membrane protein that contains one C-type lectin domain and an EGF-like domain. The gene encoding CLEC-14A maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer′s disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.Specifications
| CLEC14A | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q86T13, Q8VCP9 | |
| 161198, 66864 | |
| CLEC14A Fusion Protein Ag21081 | |
| Primary | |
| -20°C | |
| CLEC14A |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C14orf27, CEG1, CLEC14A, EGFR 5, EGFR5 | |
| Clec14a | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title