missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC14A Antibody (036), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90521H
This item is not returnable.
View return policy
Description
CLEC14A Monoclonal specifically detects CLEC14A in Mouse samples. It is validated for ELISA.
Specifications
| CLEC14A | |
| Monoclonal | |
| HRP | |
| C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGFR5, EGFR-5, Epidermal growth factor receptor 5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CLEC14A (NP_080085.3; Met1-Thr386). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse CLEC3B Mouse GL1 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 36 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 161198 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction