missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC12B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571556
This item is not returnable.
View return policy
Description
CLEC12B Polyclonal Antibody for Western Blot, IHC (P)
Cell surface receptor that protects target cells against natural killer cell-mediated lysis. Modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases.
Specifications
| CLEC12B | |
| Polyclonal | |
| Unconjugated | |
| CLEC12B | |
| 4933425B16Rik; Clec12b; C-type lectin domain family 12 member B; C-type lectin domain family 12, member B; Macrophage antigen H; RGD1560487; testis secretory sperm-binding protein Li 205a; UNQ5782; UNQ5782/PRO16089 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 387837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q2HXU8 | |
| CLEC12B | |
| KLH conjugated synthetic peptide between 249-276 amino acids from the C-terminal region of human CLEC12B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction