missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC11A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576151
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
Specifications
| CLEC11A | |
| Polyclonal | |
| Unconjugated | |
| CLEC11A | |
| AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF ß; SCGFb; SCGFbeta; SCGFß; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; ß SCGF; ßSCGF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20256, 6320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O88200, Q9Y240 | |
| CLEC11A | |
| Recombinant fusion protein of human CLEC11A. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction