missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CLEC11A Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Brand:  PeproTech 500-P99GBT-50UG

 View more versions of this product

Product Code. 17823153

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MGHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ PSASPHPLSP DQPNGGTLEN CVAQASDDGS WWDHDCQRRL YYVCEFPF. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human SCGF-beta. Anti-Human SCGF-beta-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hSCGF-beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human SCGF-beta (500-P99G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hSCGF-beta. Western Blot: To detect hSCGF-beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hSCGF-beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P99GBT-1MG will be provided as 2 x 500 μg

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLEC11A
Polyclonal
Biotin
CLEC11A
AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
Goat
Antigen affinity chromatography
RUO
6320
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q9Y240
CLEC11A
E.coli-derived, 25.0 kDa Recombinant Human SCGF-beta.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.