missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC10A, Mouse anti-Human, Clone: 1E11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010462-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: VTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAVHSEMLLRVQQLVQDLKKLTCQVATLNNNGEEASTEGTCCPVNWVEHQDSCYSpecifications
| CLEC10A | |
| Monoclonal | |
| Unconjugated | |
| C-type lectin domain family 10, member A | |
| CD301/CLECSF13/CLECSF14/HML/HML2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E11 | |
| In 1x PBS, pH 7.4 | |
| NM_006344 | |
| CLEC10A | |
| CLEC10A (NP_006335.2, 70 a.a. ∼ 169 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10462 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction