missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC10A/CD301 Antibody (ER-MP23), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64874H
This item is not returnable.
View return policy
Description
CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| CLEC10A/CD301 | |
| Monoclonal | |
| HRP | |
| CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL | |
| Balb/c macrophage precursor cell hybrids | |
| 0.125 mL | |
| Immunology | |
| 10462 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| ER-MP23 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction