missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC-2/CLEC-1B Antibody (AYP1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50166AF750
This item is not returnable.
View return policy
Description
CLEC-2/CLEC-1B Monoclonal specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CLEC-2/CLEC-1B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CLEC1B | |
| A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein | |
| 0.1 mL | |
| 51266 | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| AYP1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| AYP1 can block the interaction of human CLEC-2 with its endogenous ligand Podoplanin. AYP1 does not cross react with mouse CLEC-2 when tested via FACS. AYP1 does not detect CLEC-2 by western blot. For WB we recommend clone AYP2. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction