missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cleavage stimulation factor 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57530
This item is not returnable.
View return policy
Description
Cleavage stimulation factor 2 Polyclonal specifically detects Cleavage stimulation factor 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Cleavage stimulation factor 2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| betaCstF-64 variant 2, CF-1 64 kDa subunit, Cleavage stimulation factor 64 kDa subunit, cleavage stimulation factor subunit 2, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, CSTF 64 kDa subunit, CstF-64 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1478 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_001316.1 | |
| CSTF2 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa using the numbering given in entry NP_001316.1 (GeneID 1478). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction