missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Tego 2000 Disinfectant

Click to view available options
Volume (Metric):
50 mL
900 mL
5 L
20 L
Sterility:
Non-sterile
Sterile
Model:
RTU VH25S
SC VH25
SS VH25S
VH25
Quantity:
1 Ea.
4/Cs.
50 Pk.
6 x 900 mL
Unit Size:
20 L
Pack of 4
Pack of 6
Description
- Hard surface disinfectant based on amphoretics
- Supplied with certificates of analysis, sterility, and irradiation where applicable
Specifications
Specifications
| pH | ∼7 |
| Sterility | Non-sterile |
| Filtered | Unfiltered |
| Model | VH25 |
| Quantity | 1 Ea. |
| Volume (Metric) | 20 L |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/physician.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction