missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Plus Detergent
BIOPHARMA

Product Code. 15330318
Change view
Click to view available options
Volume (Metric):
5 L
20 L
Unit Size:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15330318 20 L 20 L
15340318 5 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15330318 Supplier Diversey™ Supplier No. 7513376
  • ONLINE EXCLUSIVE PRICE
    £118.80
    £98.33 / 20 L
    Save £20.47
    17% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleans in general purpose applications within various industries, including pharmaceutical and cosmetic. Diversey ClearKlens Plus Detergent is a concentrated, neutral, high-foaming liquid detergent.

  • Concentrated, neutral, high-foaming liquid detergent
  • Designed for general purpose applications
  • Rinse after use to remove solutions from product contact surfaces

Specifications

Form Liquid
pH ∼6
Sterility Non-sterile
Concentration 0.1 to 1%
Container Type Canister
Filtered Unfiltered
Volume (Metric) 20 L
compliance-icons
Product Identifier
  • ClearKlens Plus VH5
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: sodium alkylbenzenesulphonate, alcoholsC10-16, sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.