missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Multi VH46 Detergent
BIOPHARMA

Product Code. 17653085
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
This item is not returnable. View return policy
This item is not returnable. View return policy

ClearKlens Multi is a highly concentrated system based on organic solvents and surfactants used for a wide range of applications in the Pharmaceutical, Cosmetic, Food, and Beverage industries.

ClearKlens Multi is a strong detergent that can be used as is or as an additive in combination with strong alkaline cleaners to remove hard to detach coatings, film soils, or labels. ClearKlens Multi can be used for manual spraying application.

Key properties

  • Suitable for organic soil like emulsions, strong fatty formulations, coatings, also titanium dioxide based solutions and for label removal
  • Low foam properties and excellently removes persistent organic soil
  • Can be used as an auxiliary additive to alkaline cleaning solutions to boost cleaning performance and soil removal in cleaning and soaking applications
  • Excellent dirt‐carrying capacity
  • Reduced product entrainment due to good rinsing properties
  • Economy and performance: can be used in various cleaning and washing applications including CIP
  • Enhances effectiveness of cleaning operations, improving productivity, and quality

Specifications

Form Liquid
pH >11
Sterility Non-sterile
Composition Highly concentrated system based on organic solvents and surfactants
Dimensions (L x W x H) 290 x 240 x 395 mm
Volume (Metric) 20 L
compliance-icons
Product Identifier
  • ClearKlens Multi VH46 20L Surfactant
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, 2-aminoethanol, potassium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.