missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Eco Detergent
Click to view available options
pH:
∼2
Unit Size:
20 L
Description
- Defoamed organic acid based liquid detergent descaler for CIP applications
- Phosphorus and nitrogen free
- Shelf life of 2 years
Specifications
Specifications
| Product Type | Detergent |
| Form | Liquid |
| pH | ∼2 |
| Sterility | Non-sterile |
| Container Type | Canister |
| Content And Storage | Store at Room Temperature |
| Filtered | Unfiltered |
| Volume (Metric) | 20 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction