missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Deogen Detergent
BIOPHARMA

Product Code. 15310328
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
This item is not returnable. View return policy

Product Code. 15310328

Brand: Diversey™ 7522374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleans and disinfects for CIP and spray washing use. Diversey ClearKlens Deogen Detergent is a low foaming, chlorinated alkaline detergent.

  • Chlorinated alkaline detergent and disinfectant
  • Used for CIP and spray washing in pharmaceutical, cosmetic, and other industries

Specifications

Product Type Detergent
Form Liquid
pH >12
Sterility Non-sterile
Container Type Canister
Filtered Unfiltered
Volume (Metric) 20 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.