missing translation for 'onlineSavingsMsg'
Learn More

Diversey ClearKlens Cleansinald SC VH9

Product Code. 17813115
Change view
Click to view available options
Unit Size:
1.25 L
10 L
Certifications/Compliance:
For GB-United Kingdom, IE-Ireland, PL-Poland, PT-Portugal, RU-Russian Federation
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Certifications/Compliance
17813115 10 L -
17863105 1.25 L For GB-United Kingdom, IE-Ireland, PL-Poland, PT-Portugal, RU-Russian Federation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17813115 Supplier Diversey Supplier No. 101107602
  • ONLINE EXCLUSIVE PRICE
    £172.00
    £137.87 / 10 L
    Save £34.13
    19% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Concentrated disinfectant used for the cleaning and disinfection of walls, floors and work surfaces and in the Life Sciences industry.

  • Used for the cleaning and disinfection of walls, floors and work surfaces and in the Life Sciences industry
  • This is a double bagged disinfectant, which has been filtered to 0.2um and sterile filled
  • Has a broad spectrum of activity
  • Can be applied as a spray, wiping or wet sweeping without rinse
  • Small items or dismantled equipment may be immersed in a ClearKlens Cleansinald SC solution
  • Broad spectrum of activity
  • Delivered with conformity certificates

Specifications

Product Type Disinfectant Concentrate
Color-coded No
Form Concentrated Liquid
pH >12
Scent Unscented
Sterility Non-sterile
Appearance Clear Pale Yellow
Autoclavable Not Autoclavable
Color Clear
Density Relative density (g/cm3; 20°C): 1.04-1.06
Disposable Single-use
Filtered Non-filtered
Flammable No
Volume (Metric) 2 x 5 L
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ClearKlens Cleansinald VH9 2x5L Surface disinfectant
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: alkyl (C12-16) dimethylbenzyl ammonium chloride, tetrasodiumethylene diamine tetraacetate, alkyl al
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.