missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Clearklens Bi-Spore Disinfectant
BIOPHARMA

Product Code. p-6830023
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 15330308

Brand: Diversey™ 7514852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Disinfects hard surfaces with a sporicidal solution. Diversey Clearklens Bi-Sore Disinfectant is a chlorine dioxide generating system. It is effective against all types of micro-organisms.

  • Effective on hard surfaces against all types of micro-organisms, including bacteria, yeasts, fungi, spores, and viruses
  • Aseptically filled, two-part system for the creation of chlorine dioxide presented as an activator and base concentrate solutions
  • Double bagged to avoid environmental contamination

Specifications

Sterility Sterile
Filtered Filtered
Includes Base and activator
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • 5L Sterile hard surface sporicidal disinfectant
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Supplemental information
  • MIXTURE LIST-Contains : sodium chlorite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.