missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSND, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00007809-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MADEKTFRIGFIVLGLFLLALGTFLMSHDRPQVYGTFYAMGSVMVIGGIIWSMCQCYPKITFVPADSDFQGILSPKAMGLLENGLAAEMKSPSPQPPYVRLWEEAAYDQSLPDFSHIQMKVMSYSEDHRSLLAPEMGQPKLGTSDGGEGGPGDVQAWMEAAVVIHKGSDESEGERRLTQSWPGPLACPQGPAPLASFQDDLDMDSSEGSSPNASPHDREEACSPQQEPQGCRCPLDRFQDFALIDAPTLEDEPQEGQQWEIALPNNWQRYPRTKVEEKEASDTGGEEPEKEEEDLYYGLPDGAGDLLPDKELGFEPDTQGSpecifications
| BSND | |
| Polyclonal | |
| No additive | |
| NM_057176.2 | |
| BSND | |
| BSND (NP_476517.1, 1 a.a. ∼ 320 a.a) full-length human protein. | |
| RUO | |
| 7809 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Bartter syndrome, infantile, with sensorineural deafness (Barttin) | |
| BART/MGC119283/MGC119284/MGC119285 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction