missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
BIOPHARMA

Product Code. 10479382
Change view
Click to view available options
Surface Treatment:
Immulon 1 B
Immulon 2 HB
MaxiSorp
PolySorp
Unit Size:
Pack of 50
Pack of 60
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Unit Size
10479382 PolySorp Pack of 60
10635577 Immulon 1 B Pack of 50
10111005 Immulon 2 HB Pack of 50
10450343 MaxiSorp Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.
Product Code. 10479382 Supplier Thermo Scientific™ Supplier No. 475434
  • ONLINE EXCLUSIVE PRICE
    £163.77
    £133.87 / Pack of 60
    Save £29.90
    18% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of a variety of surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp and Immulon 1B are for adsorption of hydrophobic molecules.
  • Thermo Scientific Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.

Ordering Information:

Please note that availability may vary by country.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophobic
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Round
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Lipids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.