missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Clear Polytetrafluoroethylene Silicone Snap Cap

Product Code. 12367099
Click to view available options
Color:
Clear
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12367099

Brand: JG Finneran Associates™ 525011

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Diameter (Metric) Cap 11 mm
Color Clear
Material PTFE
For Use With (Application) Used on 12x32mm Snap Seal™ vials.
Quantity 1000 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.