missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer Cleanroom Port Disk Threaded Fitting, 38-400 1 in. Thread

Product Code. 30053595 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Natural Kynar™ Flex
Polypropylene
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Seal) unitSize
30053595 Natural Kynar™ Flex Pack of 10
30053545 Polypropylene Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30053595 Supplier Cole-Parmer Supplier No. 5011689
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Weld to single-use bags to transfer viscose fluids and powders

Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Wide 1 in. opening—ideal for powders and high viscosity fluid

Specifications

Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 1 in.
Packaging Quantity 10/Pk.
Outlet Port Port Disk x Thread
Sterilization Method 121°C
Shape Straight
Material (Seal) Natural Kynar™ Flex
Color Natural

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.