Learn More
Cole-Parmer Cleanroom Port Disk Hose Barb Fittings, Mini Dome, Engage™ 8402
Product Code. p-200105021 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/2 in.
1/4 in.
1/8 in.
3/26 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
Product Code. 30053833
Brand: Cole-Parmer 5011675
Only null left
Add to basket
Add to basket
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Mini-dome protrusions inhibit suction blockage. Ultrasonic, radio frequency, or thermal weld to polyethylene bags. Meets FDA requirements 21 CFR 177.1520(c) 3.2c and 21 CFR 176.170(c) and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/4 in.
Packaging Quantity 10/Pk.
Outlet Port Port Disk x Hose Barb
Sterilization Method Gamma irradiation up to 50 kGy
Shape Straight
Material (Seal) Engage™ 8402
Color Translucent