Learn More
Cole-Parmer Cleanroom Large-Bore Male Luer to Hose Barb Fittings, Straight Adapter, Natural Nylon
Product Code. p-200105157 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/4 in.
3/16 in.
5/16 in.
Unit Size:
Pack of 10
Product Code. 30053238
Brand: Cole-Parmer 5010964
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. RoHS compliant
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 5/16 in.
Material Nylon
Packaging Quantity 10/Pk.
Inlet Large Bore Male Luer, Hose Barb
Color Natural
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.