Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, Natural Kynar™
Product Code. p-200105118 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
Product Code. 30054263
Brand: Cole-Parmer 5011143
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Certifications/Compliance USP;Non-DEHP;Non-Phthalate
Diameter (English) Inner 5/16 in.
Min. Operating Temperature (Metric) -35°C
Packaging Quantity 10/Pk.
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Shape Y
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.