Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Tee Union, High-Density Polyethylene
Product Code. 30053904 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/4 in.
3/8 in.
Unit Size:
Pack of 10
Product Code. 30053904
Brand: Cole-Parmer 5011110
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520 (when used unmodified for the manufacture of food contact articles) and RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/8 in.
Packaging Quantity 10/Pk.
Outlet Port Hose Barb Tee
Shape Tee
Color White